Stampa…

D-(-)-Valine ≥98.0% (by titrimetric analysis)

Fornitore: TCI

Synonyms: H-D-Val-OH, (R)-(-)-Valine

V0012-1G V0012-25G
TCIAV0012-1GEA 16 EUR
TCIAV0012-1G TCIAV0012-25G
D-(-)-Valine ≥98.0% (by titrimetric analysis)
D-(-)-Valine
Formula: C₅H₁₁NO₂
Punto di ebollizione: 217 °C (1013 hPa)
Punto di fusione: 86 °C
MDL Number: MFCD00064219
CAS Number: 640-68-6
EINECS: 211-368-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=694d68472bcd090d82791d226a4df621200746f0&vid=694d68472bcd090d82791d226a4df621200746f0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Optical purity (LC) min. 99.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -26.0 to -29.0 deg (C=8, 6mol/L HCl)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR