(S)-(+)-2-Amino-1-propanol ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
L(+)-Alaninol, L-Alaninol, (S)-2-Aminopropan-1-ol, L(+)-2-Amino-1-propanol
A1085-1ML
A1085-25ML
A1085-5ML
TCIAA1085-1MLEA
16
EUR
TCIAA1085-1ML
TCIAA1085-25ML
TCIAA1085-5ML
(S)-(+)-2-Amino-1-propanol ≥98.0% (by GC, titration analysis)
(S)-(+)-2-Amino-1-propanol
Formula:
C₃H₉NO Peso molecolare: 75,11 g/mol Punto di ebollizione: 98°C Densità: 0,955 g/cm³ (20 °C) Flash Pt: 83°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00064412 CAS Number: 2749-11-3 EINECS: 220-388-7 UN: 2735 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +21.0 to +23.0 deg (C=2, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...