Stampa…

Semicarbazide Idrocloruro ≥99.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

Synonyms: Semicarbazide hydrochloride, Carbamylhydrazine hydrochloride, Aminourea hydrochloride, Hydrazinecarboxamide hydrochloride, N-Aminourea hydrochloride, Hydrazinecarboxamidehydrochloride

S0032-25G S0032-500G
TCIAS0032-25GEA 16 EUR
TCIAS0032-25G TCIAS0032-500G
Semicarbazide Idrocloruro ≥99.0% (by titrimetric analysis)
Semicarbazide Idrocloruro
Formula: CH₆ClN₃O
Peso molecolare: 111.53 g/mol
MDL Number: MFCD00013009
CAS Number: 563-41-7
EINECS: 209-247-0
UN: 2811
ADR: 6.1 III
Merck Index: 14,08444
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d782b35d8db77b8d65f90581338eaa81537c1c32&vid=d782b35d8db77b8d65f90581338eaa81537c1c32&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Bromination Method) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR