Stampa…

(±)-Phenylsuccinic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

P0155-25G P0155-500G
TCIAP0155-25GEA 18 EUR
TCIAP0155-25G TCIAP0155-500G
(±)-Phenylsuccinic acid ≥98.0% (by HPLC, titration analysis)
(±)-Phenylsuccinic acid
Formula: C₁₀H₁₀O₄
Punto di ebollizione: 368 °C (1013 hPa)
Punto di fusione: 167…169 °C
MDL Number: MFCD00004256
CAS Number: 635-51-8
EINECS: 211-238-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad79865bc2fa4a49b7cbf85a8d6adb5a97f4ea7b&vid=ad79865bc2fa4a49b7cbf85a8d6adb5a97f4ea7b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR