DL-m-Tyrosine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Hydroxyphenylalanine, 3-(m-Hidroxifenil)-DL-alanina, 3-(3-Hydroxyphenyl)-DL-alanine
T0787-1G
T0787-5G
TCIAT0787-1GEA
35
EUR
TCIAT0787-1G
TCIAT0787-5G
DL-m-Tyrosine ≥98.0% (by HPLC, titration analysis)
DL-m-Tyrosine
Formula:
C₉H₁₁NO₃ Peso molecolare: 181,19 g/mol Punto di ebollizione: 351 °C (1013 hPa) Punto di fusione: 283 °C Densità: 1,456 g/cm³ (20 °C) |
MDL Number:
MFCD00002597 CAS Number: 775-06-4 EINECS: 212-270-9 Merck Index: 14,09839 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in 1mol/L HCl | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...