Stampa…

4-Nitrocinnamic acid ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(4-Nitrophenyl)propenoic acid, 3-(4-Nitrophenyl)acrylic acid

N0355-10G N0355-25G
TCIAN0355-10GEA 45 EUR
TCIAN0355-10G TCIAN0355-25G
4-Nitrocinnamic acid ≥99.0% (by titrimetric analysis)
4-Nitrocinnamic acid
Formula: C₉H₇NO₄
Peso molecolare: 193,16 g/mol
MDL Number: MFCD00007381
CAS Number: 619-89-6
EINECS: 210-617-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4681eb455cd9be310ec3847e4469736bafd11c4&vid=b9d81111fb9cabe8acf994fca10681defe5e0175&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 99.0 %
Solubility in 1mol/L NaOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR