Stampa…

4-Chlorocinnamic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(4-Chlorophenyl)acrylic acid

C0149-25G C0149-500G
TCIAC0149-25GEA 42 EUR
TCIAC0149-25G TCIAC0149-500G
4-Chlorocinnamic acid ≥98.0% (by GC, titration analysis)
4-Chlorocinnamic acid
Formula: C₉H₇ClO₂
Peso molecolare: 182,61 g/mol
Punto di ebollizione: 311 °C (1013 hPa)
Punto di fusione: 252…254 °C
MDL Number: MFCD00004396
CAS Number: 1615-02-7
EINECS: 216-564-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a43b81efbe516bafc0a1d4148dc13c47b9873abb&vid=615eede5c0d33da0c45b93b9d474f07225d09909&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR