Stampa…

3-Fluorocinnamic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(3-Fluorophenyl)acrylic acid, m-Fluorocinnamic acid, 3-Fluorocinnamic Acid

F0264-25G F0264-5G
TCIAF0264-25GEA 116 EUR
TCIAF0264-25G TCIAF0264-5G
3-Fluorocinnamic acid ≥98.0% (by titrimetric analysis)
3-Fluorocinnamic acid
Formula: C₉H₇FO₂
Peso molecolare: 166,15 g/mol
MDL Number: MFCD00004383
CAS Number: 458-46-8
EINECS: 207-281-0
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c854c9e935375bc4622423b17367bda8c7515f6&vid=1c854c9e935375bc4622423b17367bda8c7515f6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 165.0 to 168.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR