Stampa…

Ethyl thiocyanate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Ethyl Rhodanide, Thiocyanic acid ethyl ester

T0200-25ML T0200-500ML
TCIAT0200-25MLEA 166 EUR
TCIAT0200-25ML TCIAT0200-500ML
Ethyl thiocyanate ≥98.0% (by GC)
Ethyl thiocyanate
Formula: C₃H₅NS
Punto di ebollizione: 145°C
Punto di fusione: –86 °C
Densità: 1,01 g/cm³ (20 °C)
Flash Pt: 42°C
MDL Number: MFCD00001834
CAS Number: 542-90-5
EINECS: 208-833-3
UN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61ea27fb839ed49080d046787b98a36e5a879da3&vid=61ea27fb839ed49080d046787b98a36e5a879da3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR