Stampa…

Tetrapropylammonium bromide ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

T0694-25G T0694-500G
TCIAT0694-25GEA 16 EUR
TCIAT0694-25G TCIAT0694-500G
Tetrapropylammonium bromide ≥98.0% (by titrimetric analysis)
Tetrapropylammonium bromide
Formula: C₁₂H₂₈BrN
Punto di fusione: 265…274 °C
MDL Number: MFCD00011840
CAS Number: 1941-30-6
EINECS: 217-727-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=432cb826b9a3513ab83a5cfc436b91e3ad02d9c2&vid=432cb826b9a3513ab83a5cfc436b91e3ad02d9c2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR