Stampa…

Ethyl myristate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Tetradecanoic acid ethyl ester, Tetradecylic acid ethyl ester, Ethyl tetradecanoate, Myristic acid ethyl ester

M0479-25ML M0479-500ML M0479-100ML
TCIAM0479-25MLEA 16 EUR
TCIAM0479-25ML TCIAM0479-500ML TCIAM0479-100ML
Ethyl myristate ≥98.0% (by GC)
Ethyl myristate
Formula: C₁₆H₃₂O₂
Peso molecolare: 256,43 g/mol
Punto di ebollizione: 309°C
Densità: 0,86 g/cm³ (20 °C)
Flash Pt: 167°C
MDL Number: MFCD00008984
CAS Number: 124-06-1
EINECS: 204-675-4
Merck Index: 14,06333
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8aaee6c6f8ab93955479e7d733cb3864dfacd6b&vid=f8aaee6c6f8ab93955479e7d733cb3864dfacd6b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR