Stampa…

Tetrabutylphosphonium bromide ≥99.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

T1124-25G T1124-500G T1124-100G
TCIAT1124-25GEA 22 EUR
TCIAT1124-25G TCIAT1124-500G TCIAT1124-100G
Tetrabutylphosphonium bromide ≥99.0% (by titrimetric analysis)
Tetrabutylphosphonium bromide
Formula: C₁₆H₃₆BrP
Punto di fusione: 95…101 °C
MDL Number: MFCD00011853
CAS Number: 3115-68-2
EINECS: 221-487-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6929090fdcfed1067fdc5cbc8c9ba3d9a7a2fb7&vid=d6929090fdcfed1067fdc5cbc8c9ba3d9a7a2fb7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 99.0 %
Melting point 103.0 to 107.0 deg-C
Solubility in Water almost transparency

Specifiche

Environmentally Preferable Più sicuro dal punto di vista della salute umana e dell'ambiente

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR