Stampa…

Tween® 85 (Polysorbate)

Fornitore: TCI

Synonyms: Polyoxyethylenesorbitan Trioleate

T0547-25G T0547-500G
TCIAT0547-25GEA 17 EUR
TCIAT0547-25G TCIAT0547-500G
Tween® 85 (Polysorbate)
Tween® 85 (Polysorbate)
Flash Pt: 315°C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01779641
CAS Number: 9005-70-3

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown clear liquid to cloudy liquid
Saponification Value 83.0 to 105.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR