Stampa…

Octadecylcyclohexane ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Stearylcyclohexane, 1-Cyclohexyloctadecane

O0161-25G O0161-5G
TCIAO0161-25GEA 532 EUR
TCIAO0161-25G TCIAO0161-5G
Octadecylcyclohexane ≥98.0% (by GC)
Octadecylcyclohexane
Formula: C₂₄H₄₈
Peso molecolare: 336,65 g/mol
Punto di ebollizione: 409°C
MDL Number: MFCD00058953
CAS Number: 4445-06-1
EINECS: 224-683-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cb3ea2c2a18b05d731a0ff2108a30bc3e64bc92&vid=0cb3ea2c2a18b05d731a0ff2108a30bc3e64bc92&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 44.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR