Stampa…

7-Bromoindole ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 7-Bromo-1H-indole

B2873-1G B2873-5G
TCIAB2873-1GEA 37 EUR
TCIAB2873-1G TCIAB2873-5G
7-Bromoindole ≥97.0% (by GC)
7-Bromoindole
Formula: C₈H₆BrN
Peso molecolare: 196,05 g/mol
MDL Number: MFCD00799492
CAS Number: 51417-51-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abcc7de6b5a98ea1b21d44bd411462be30b154c2&vid=abcc7de6b5a98ea1b21d44bd411462be30b154c2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 44.0 to 48.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR