6-Fluoroindole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Fluoro-1H-indole
F0352-1G
F0352-5G
TCIAF0352-1GEA
35
EUR
TCIAF0352-1G
TCIAF0352-5G
6-Fluoroindole ≥98.0% (by GC)
6-Fluoroindole
Formula:
C₈H₆FN Peso molecolare: 135,14 g/mol Punto di fusione: 72…77 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00056933 CAS Number: 399-51-9 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 78.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...