6-Aminohexan-1-ol ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Amino-1-hexanol
A1027-25G
A1027-5G
TCIAA1027-25GEA
135
EUR
TCIAA1027-25G
TCIAA1027-5G
6-Aminohexan-1-ol ≥97.0% (by GC, titration analysis)
6-Aminohexan-1-ol
Formula:
C₆H₁₅NO Peso molecolare: 117,19 g/mol Punto di ebollizione: 225°C Punto di fusione: 53…55 °C |
MDL Number:
MFCD00008241 CAS Number: 4048-33-3 EINECS: 223-748-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 54.0 to 59.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...