Stampa…

5-Idrossiindolo ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Idrossi-1H-indolo

H0252-1G H0252-100MG H0252-5G
TCIAH0252-1GEA 30 EUR
TCIAH0252-1G TCIAH0252-100MG TCIAH0252-5G
5-Idrossiindolo ≥98.0% (by GC)
5-Idrossiindolo
Formula: C₈H₇NO
Peso molecolare: 133,15 g/mol
Punto di fusione: 107…108 °C
MDL Number: MFCD00005677
CAS Number: 1953-54-4
EINECS: 217-782-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1b6fafce0d528a5669e45ea9a3ad7847a84492a&vid=b1b6fafce0d528a5669e45ea9a3ad7847a84492a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 110.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR