Stampa…

4-Phenylbutyric acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 4-Phenylbutanoic acid

P0643-25G P0643-250G
TCIAP0643-25GEA 38 EUR
TCIAP0643-25G TCIAP0643-250G
4-Phenylbutyric acid ≥98.0% (by GC, titration analysis)
4-Phenylbutyric acid
Formula: C₁₀H₁₂O₂
Punto di ebollizione: 175°C
Punto di fusione: 48…51 °C
MDL Number: MFCD00004403
CAS Number: 1821-12-1
EINECS: 217-341-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d593e3c252db6e92e8166d558ca31ae679f03a15&vid=d593e3c252db6e92e8166d558ca31ae679f03a15&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 49.0 to 53.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR