4-Nitrobenzoil cloruro ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
p-Nitrobenzoyl chloride
Formula:
C₇H₄ClNO₃ Punto di ebollizione: 128°C Punto di fusione: 97…98 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007345 CAS Number: 122-04-3 EINECS: 204-517-4 UN: 3261 ADR: 8,II Merck Index: 14,06589 |
|
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 72.0 to 75.0 deg-C |
Solubility in Toluene | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...