4-Hydroxycoumarin ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
H0235-25G
H0235-250G
TCIAH0235-25GEA
30
EUR
TCIAH0235-25G
TCIAH0235-250G
4-Hydroxycoumarin ≥98.0% (by GC, titration analysis)
4-Hydroxycoumarin
Formula:
C₉H₆O₃ Punto di ebollizione: 349 °C (1013 hPa) Punto di fusione: 211…214 °C |
MDL Number:
MFCD00006856 CAS Number: 1076-38-6 EINECS: 214-060-2 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...