4-Fluoro-o-toluic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
F0559-25G
F0559-5G
TCIAF0559-25GEA
295
EUR
TCIAF0559-25G
TCIAF0559-5G
4-Fluoro-o-toluic acid ≥98.0% (by GC, titration analysis)
4-Fluoro-o-toluic acid
Formula:
C₈H₇FO₂ Peso molecolare: 154,14 g/mol |
MDL Number:
MFCD04115880 CAS Number: 321-21-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 172.0 to 176.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...