Stampa…

4-Aminobenzamide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

A0631-25G A0631-500G
TCIAA0631-25GEA 17 EUR
TCIAA0631-25G TCIAA0631-500G
4-Aminobenzamide ≥98.0% (by HPLC, titration analysis)
4-Aminobenzamide
Formula: C₇H₈N₂O
Peso molecolare: 136,15 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007999
CAS Number: 2835-68-9
EINECS: 220-612-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6c16667a05f7894765e93848195ef3938d1d3d8&vid=f6c16667a05f7894765e93848195ef3938d1d3d8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 181.0 to 185.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR