4-Amino-3-nitropyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine
A2260-25G
A2260-5G
TCIAA2260-25GEA
236
EUR
TCIAA2260-25G
TCIAA2260-5G
4-Amino-3-nitropyridine ≥98.0% (by GC, titration analysis)
4-Amino-3-nitropyridine
Formula:
C₅H₅N₃O₂ Peso molecolare: 139,11 g/mol |
MDL Number:
MFCD00160304 CAS Number: 1681-37-4 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 203.0 to 207.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...