Acido 4-indolil-3-butirrico ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
3-Indolebutyric acid, Indole-3-butyric acid
Formula:
C₁₂H₁₃NO₂ Peso molecolare: 203.24 g/mol Densità: 1.27 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00005664 CAS Number: 133-32-4 EINECS: 205-101-5 UN: 2811 ADR: 6.1 III Merck Index: 14,04965 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 123.0 to 126.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...