Stampa…

Rosso bromofenolo ≥80.0% (by HPLC)

Fornitore: TCI

Synonyms: 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromophenol] S,S-dioxide

B0632-1G B0632-25G
TCIAB0632-1GEA 42 EUR
TCIAB0632-1G TCIAB0632-25G
Rosso bromofenolo ≥80.0% (by HPLC)
Rosso bromofenolo
Formula: C₁₉H₁₂Br₂O₅S
Peso molecolare: 512.18 g/mol
Punto di fusione: 230
MDL Number: MFCD00023014
CAS Number: 2800-80-8
EINECS: 220-538-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fdac9ed5a95402f60eab62051431bd599c30edc&vid=9fdac9ed5a95402f60eab62051431bd599c30edc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Dark red to Brown powder to crystal
Lambda max. 574.0 to 577.0 nm (pH 6.8 buffer sol.)
Absorbance min. 0.560 (5mg/L, pH 6.8 buffer sol., 574.0 to 577.0 nm)
Transition interval (pH) (redish yellow)5.2 to 6.8 (red purple)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR