Stampa…

3-Chloro-2-hydroxypyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Chloro-2-pyridinol

C2340-1G C2340-5G
TCIAC2340-1GEA 26 EUR
TCIAC2340-1G TCIAC2340-5G
3-Chloro-2-hydroxypyridine ≥98.0% (by GC)
3-Chloro-2-hydroxypyridine
Formula: C₅H₄ClNO
Peso molecolare: 129,55 g/mol
MDL Number: MFCD01646109
CAS Number: 13466-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a981d4ea700eb2d5935cc7bb4f76506868fb0cd&vid=2a981d4ea700eb2d5935cc7bb4f76506868fb0cd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 174.0 to 180.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR