5-Bromo-2-methoxypyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
B2398-25G
B2398-5G
TCIAB2398-25GEA
77
EUR
TCIAB2398-25G
TCIAB2398-5G
5-Bromo-2-methoxypyridine ≥98.0% (by GC, titration analysis)
5-Bromo-2-methoxypyridine
Formula:
C₆H₆BrNO Peso molecolare: 188.02 g/mol Punto di ebollizione: 195°C Flash Pt: 96°C |
MDL Number:
MFCD01318952 CAS Number: 13472-85-0 |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...