Stampa…

3-Bromo-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

B3370-1G B3370-5G
TCIAB3370-1GEA 36 EUR
TCIAB3370-1G TCIAB3370-5G
3-Bromo-5-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-5-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MDL Number: MFCD00100098
CAS Number: 6307-83-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19dba17a4f8a60d82e5811bd3e124cf466740f24&vid=19dba17a4f8a60d82e5811bd3e124cf466740f24&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 164.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR