3-Bromo-5-chloropyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B3608-1G
B3608-5G
TCIAB3608-1GEA
38
EUR
TCIAB3608-1G
TCIAB3608-5G
3-Bromo-5-chloropyridine ≥98.0% (by GC)
3-Bromo-5-chloropyridine
Formula:
C₅H₃BrClN Peso molecolare: 192,44 g/mol |
MDL Number:
MFCD04114221 CAS Number: 73583-39-8 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...