Ethyl-3-aminobenzoate ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₉H₁₁NO₂ Punto di ebollizione: 294°C Densità: 1,173 g/cm³ (20 °C) |
MDL Number:
MFCD00007794 CAS Number: 582-33-2 EINECS: 209-482-9 |
|
Risultati del test di specificazione
Appearance | White or Colorless to Light orange to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...