Stampa…

3-Amino-4-methylpyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

A1957-25G A1957-5G
TCIAA1957-25GEA 136 EUR
TCIAA1957-25G TCIAA1957-5G
3-Amino-4-methylpyridine ≥98.0% (by GC, titration analysis)
3-Amino-4-methylpyridine
Formula: C₆H₈N₂
Peso molecolare: 108,14 g/mol
Punto di ebollizione: 254°C
MDL Number: MFCD00128871
CAS Number: 3430-27-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f530fc81f55a6abb548d1e5b304270141f289642&vid=f530fc81f55a6abb548d1e5b304270141f289642&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 103.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR