Stampa…

3-Amino-2-bromopyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Bromopyridin-3-amine, 2-Bromo-3-pyridylamine

A1898-1G A1898-5G
TCIAA1898-1GEA 24 EUR
TCIAA1898-1G TCIAA1898-5G
3-Amino-2-bromopyridine ≥98.0% (by GC, titration analysis)
3-Amino-2-bromopyridine
Formula: C₅H₅BrN₂
Peso molecolare: 173,01 g/mol
MDL Number: MFCD00234064
CAS Number: 39856-58-1
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=935f36be262e42944c96bbbd59fdfa89a31ad10e&vid=935f36be262e42944c96bbbd59fdfa89a31ad10e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 77.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR