3,5-Difluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
D3407-25G
D3407-5G
TCIAD3407-25GEA
113
EUR
TCIAD3407-25G
TCIAD3407-5G
3,5-Difluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
3,5-Difluorobenzoyl chloride
Formula:
C₇H₃ClF₂O Peso molecolare: 176,55 g/mol Densità: 1,5 g/cm³ (20 °C) Flash Pt: 69°C |
MDL Number:
MFCD00010309 CAS Number: 129714-97-2 UN: 3265 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...