2-Fluoro-5-iodopyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
F0523-1G
F0523-5G
TCIAF0523-1GEA
24
EUR
TCIAF0523-1G
TCIAF0523-5G
2-Fluoro-5-iodopyridine ≥98.0% (by GC)
2-Fluoro-5-iodopyridine
Formula:
C₅H₃FIN Peso molecolare: 222,99 g/mol |
MDL Number:
MFCD03095301 CAS Number: 171197-80-1 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 32.0 to 37.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...