Stampa…
2-Chloro-5-iodopyridine ≥98.0% (by GC)
Fornitore: TCI
Attenzione
C2341-5G C2341-25G
TCIAC2341-5GEA 61 EUR
TCIAC2341-5G TCIAC2341-25G
2-Chloro-5-iodopyridine ≥98.0% (by GC)
2-Chloro-5-iodopyridine
Formula: C₅H₃ClIN
Peso molecolare: 239.44 g/mol
MDL Number: MFCD01863635
CAS Number: 69045-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=013cd4a979a8c0cf86402a2581d25952eeabddd4&vid=013cd4a979a8c0cf86402a2581d25952eeabddd4&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Amber powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C