Ethyl anthranilate ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Aminobenzoic acid ethyl ester, o-Aminobenzoic acid ethyl ester, Ethyl 2-aminobenzoate
A0499-25ML
A0499-500ML
A0499-100ML
TCIAA0499-25MLEA
16
EUR
TCIAA0499-25ML
TCIAA0499-500ML
TCIAA0499-100ML
Ethyl anthranilate ≥99.0% (by GC)
Ethyl anthranilate
Formula:
C₉H₁₁NO₂ Peso molecolare: 165,19 g/mol Punto di ebollizione: 268 °C (1013 hPa) Densità: 1,12 g/cm³ (20 °C) Flash Pt: 138°C |
MDL Number:
MFCD00007711 CAS Number: 87-25-2 EINECS: 201-735-1 |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...