2-Amino-6-methylpyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
6-Methyl-2-pyridylamine, 6-Methylpyridin-2-amine, 2-Amino-6-picoline
A0403-25G
A0403-500G
A0403-100G
TCIAA0403-25GEA
17
EUR
TCIAA0403-25G
TCIAA0403-500G
TCIAA0403-100G
2-Amino-6-methylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-6-methylpyridine
Formula:
C₆H₈N₂ Peso molecolare: 108,14 g/mol Punto di ebollizione: 209°C Punto di fusione: 40…43 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00006331 CAS Number: 1824-81-3 EINECS: 217-360-1 UN: 2811 ADR: 6.1,II |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 42.0 to 45.0 deg-C |
Solubility in Water | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...