2-Amino-6-bromopyridine ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Bromo-2-pyridylamine, 6-Bromopyridin-2-amine
A1529-25G
A1529-5G
TCIAA1529-25GEA
160
EUR
TCIAA1529-25G
TCIAA1529-5G
2-Amino-6-bromopyridine ≥98.0% (by titrimetric analysis)
2-Amino-6-bromopyridine
Formula:
C₅H₅BrN₂ Peso molecolare: 173,01 g/mol |
MDL Number:
MFCD00137843 CAS Number: 19798-81-3 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Methanol | very faint turbidity |
Melting point | 87.0 to 91.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...