Stampa…

2-Amino-5-fluoropyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

A1664-1G A1664-5G
TCIAA1664-1GEA 23 EUR
TCIAA1664-1G TCIAA1664-5G
2-Amino-5-fluoropyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-fluoropyridine
Formula: C₅H₅FN₂
Peso molecolare: 112.11 g/mol
MDL Number: MFCD01861120
CAS Number: 21717-96-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1363fe35f6160a3709837f78780a74468ce034bd&vid=1363fe35f6160a3709837f78780a74468ce034bd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR