Stampa…
2-Amino-5-bromo-4-methylpyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 5-Bromo-4-methylpyridin-2-amine, 5-Bromo-4-methyl-2-pyridylamine, 2-Amino-5-bromo-4-picoline
A1897-1G A1897-5G
TCIAA1897-1GEA 16 EUR
TCIAA1897-1G TCIAA1897-5G
2-Amino-5-bromo-4-methylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-5-bromo-4-methylpyridine
Formula: C₆H₇BrN₂
Peso molecolare: 187,04 g/mol
MDL Number: MFCD03427660
CAS Number: 98198-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dddd60303b0ffe097bc83bd1315dd76f20f275c9&vid=dddd60303b0ffe097bc83bd1315dd76f20f275c9&prid=web&clid=SAPDAM
Visualizza i codici
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 148.0 to 152.0 deg-C