Teobromina ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₇H₈N₄O₂ Punto di fusione: 350 °C Densità: 1,419 g/cm³ (25 °C) |
MDL Number:
MFCD00022830 CAS Number: 83-67-0 EINECS: 201-494-2 Merck Index: 14,09282 |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...