Stampa…

4-Nitrochalcone ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Nitrobenzylidenacetophenone

N0839-25G N0839-5G
TCIAN0839-25GEA 165 EUR
TCIAN0839-25G TCIAN0839-5G
4-Nitrochalcone ≥95.0% (by GC)
4-Nitrochalcone
Formula: C₁₅H₁₁NO₃
Peso molecolare: 253,26 g/mol
Punto di fusione: 165 °C
MDL Number: MFCD00007382
CAS Number: 1222-98-6
EINECS: 214-949-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c1e05b9957231040daee236cf337f05a99d5e73&vid=8c1e05b9957231040daee236cf337f05a99d5e73&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 163.0 to 167.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR