Stampa…

3-Nitro-o-cresol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Methyl-3-nitrophenol, 1-Hydroxy-2-methyl-3-nitrobenzene

N0627-25G N0627-5G
TCIAN0627-25GEA 96 EUR
TCIAN0627-25G TCIAN0627-5G
3-Nitro-o-cresol ≥98.0% (by GC)
3-Nitro-o-cresol
Formula: C₇H₇NO₃
Peso molecolare: 153,14 g/mol
Punto di fusione: 149…152 °C
MDL Number: MFCD00007241
CAS Number: 5460-31-1
EINECS: 226-739-0
UN: 2446
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffd4881cc438070ca80cb63544811fa0e79c1069&vid=ffd4881cc438070ca80cb63544811fa0e79c1069&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR