Stampa…

4-Bromo-2-methylanisole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Bromo-2-methoxytoluene, 1-Bromo-4-methoxy-3-methylbenzene, 4-Bromo-1-methoxy-2-methylbenzene

B1910-5G B1910-25G
TCIAB1910-5GEA 32 EUR
TCIAB1910-5G TCIAB1910-25G
4-Bromo-2-methylanisole ≥98.0% (by GC)
4-Bromo-2-methylanisole
Formula: C₈H₉BrO
Peso molecolare: 201,06 g/mol
Punto di ebollizione: 124°C
MDL Number: MFCD01321139
CAS Number: 14804-31-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 70.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR