4-Bromobenzyl alcohol ≥99.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
(4-Bromophenyl)methanol
B1372-25G
B1372-100G
TCIAB1372-25GEA
40
EUR
TCIAB1372-25G
TCIAB1372-100G
4-Bromobenzyl alcohol ≥99.0% (by GC)
4-Bromobenzyl alcohol
Formula:
C₇H₇BrO Peso molecolare: 187,04 g/mol Punto di ebollizione: 268 °C (1013 hPa) Punto di fusione: 77…78 °C |
MDL Number:
MFCD00004650 CAS Number: 873-75-6 EINECS: 212-851-7 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 99.0 % |
Solubility in Methanol | very faint turbidity |
Melting point | 77.0 to 80.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...