4-Amino-3-nitrophenol ≥97.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₆H₆N₂O₃ Punto di fusione: 149…152 °C |
MDL Number:
MFCD00066310 CAS Number: 610-81-1 EINECS: 210-236-8 |
|
Risultati del test di specificazione
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 148.0 to 154.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...