Stampa…

4-Bromo-2,6-difluoroaniline ≥98.0% (by GC)

Fornitore: TCI
Attenzione

B3220-1G B3220-25G B3220-5G
TCIAB3220-1GEA 30 EUR
TCIAB3220-1G TCIAB3220-25G TCIAB3220-5G
4-Bromo-2,6-difluoroaniline ≥98.0% (by GC)
4-Bromo-2,6-difluoroaniline
Formula: C₆H₄BrF₂N
MDL Number: MFCD00013389
CAS Number: 67567-26-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebf90c030763ec63bae3175aee15e71df9960dfd&vid=ebf90c030763ec63bae3175aee15e71df9960dfd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR