Stampa…

4-Bromo-2,5-difluoroaniline ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B2072-1G B2072-5G
TCIAB2072-1GEA 17 EUR
TCIAB2072-1G TCIAB2072-5G
4-Bromo-2,5-difluoroaniline ≥98.0% (by GC)
4-Bromo-2,5-difluoroaniline
Formula: C₆H₄BrF₂N
Peso molecolare: 208,01 g/mol
MDL Number: MFCD01861125
CAS Number: 112279-60-4
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6abff8cdc567725962e2c10ced470774e05ef82c&vid=6abff8cdc567725962e2c10ced470774e05ef82c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 74.0 to 77.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR