6-Cloro-o-toluidina ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Chloro-6-methylaniline, 2-Amino-3-chlorotoluene
A1959-25G
A1959-5G
TCIAA1959-25GEA
139
EUR
TCIAA1959-25G
TCIAA1959-5G
6-Cloro-o-toluidina ≥98.0% (by GC, titration analysis)
6-Cloro-o-toluidina
Formula:
C₇H₈ClN Peso molecolare: 141,6 g/mol Punto di ebollizione: 215 °C (1013 hPa) Punto di fusione: 2 °C Densità: 1,152 g/cm³ (20 °C) |
MDL Number:
MFCD00007679 CAS Number: 87-63-8 EINECS: 201-759-2 UN: 3429 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Red clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...